2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98.0%

Supplier: TCI America

Synonyms: DavePhos, 2-(Dicyclohexylphosphino)-2'-(N,N-dimethylamino))-1,1'-biphenyl, 2'-(Dicyclohexylphosphino)-N,N-dimethyl-[1,1'-biphenyl]-2-amine, 2-Dicyclohexylphosphino-2’-(N,N-dimethylamino)biphenyl

D3389-1G D3389-5G
TCD3389-1GEA 204.93 CAD
TCD3389-1G TCD3389-5G
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl ≥98.0%
2-(Dicyclohexylphosphino)-2'-(dimethylamino)biphenyl

CAS Number: 213697-53-1
Molecular Formula: C26H36NP
Molecular Weight: 393.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 123


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Ligands
Other Product Names / Synonyms: DavePhos

Formula: C₂₆H₃₆NP
MW: 393.55 g/mol
Melting Pt: 123 °C
MDL Number: MFCD02183572
CAS Number: 213697-53-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=13f53e0d56969f336cfab01c34ecd97da1883bb2&vid=13f53e0d56969f336cfab01c34ecd97da1883bb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR