3,5-Dibromotoluene ≥98.0%

Supplier: TCI America

Synonyms: 1,3-Dibromo-5-methylbenzene

D3376-5G D3376-25G
TCD3376-5GEA 31.6 CAD
TCD3376-5G TCD3376-25G
3,5-Dibromotoluene ≥98.0%
3,5-Dibromotoluene

CAS Number: 1611-92-3
MDL Number: MFCD00013528
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 246
Melting point (°C): 41
Flash Point (°C): 113

Formula: C₇H₆Br₂
MW: 249.93
Boiling Pt: 246 °C
Melting Pt: 41 °C
Flash Pt: 113 °C
MDL Number: MFCD00013528
CAS Number: 1611-92-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d34fe65afda93372d4eab09ff27c106bcfbb30&vid=46763448cf6702a54364cbcea777c61b34902079&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR