Diethyl ethyl(phenyl)malonate ≥98.0%
Supplier: TCI America
D3375-25G
D3375-5G
TCD3375-25GEA
93.46
CAD
TCD3375-25G
TCD3375-5G
Diethyl ethyl(phenyl)malonate ≥98.0%
Diethyl ethyl(phenyl)malonate
CAS Number: 76-67-5
MDL Number: MFCD00040759
Molecular Formula: C15H20O4
Molecular Weight: 264.32
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 171
Melting point (°C): -7
Flash Point (°C): 113
Specific Gravity (20/20): 1.07
Other Product Names / Synonyms: Ethyl(phenyl)malonic Acid Diethyl Ester
Formula:
C₁₅H₂₀O₄ Boiling Pt: 171 °C Melting Pt: –7 °C Density: 1.07 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00040759 CAS Number: 76-67-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...