3',4'-Dichloropropiophenone ≥98.0%

Supplier: TCI America

D3365-25G D3365-5G
TCD3365-25GEA 447.93 CAD
TCD3365-25G TCD3365-5G
3',4'-Dichloropropiophenone ≥98.0%
3',4'-Dichloropropiophenone

CAS Number: 6582-42-9
MDL Number: MFCD00000554
Molecular Formula: C9H8Cl2O
Molecular Weight: 203.06
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 247
Melting point (°C): 46
Flash Point (°C): 113


Other Product Names / Synonyms: 3,4-Dichlorophenyl Ethyl Ketone

Formula: C₉H₈Cl₂O
Boiling Pt: 247 °C
Melting Pt: 46 °C
Flash Pt: 113 °C
MDL Number: MFCD00000554
CAS Number: 6582-42-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=be4fc3a9f590dd8854ad99108e22b5597c5c4a37&vid=be4fc3a9f590dd8854ad99108e22b5597c5c4a37&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR