Diethyl (4-methylbenzyl)phosphonate ≥98.0%

Supplier: TCI America

Synonyms: Diethyl 4-methylbenzylphosphonate

D3336-5G D3336-25G
TCD3336-5GEA 67.94 CAD
TCD3336-5G TCD3336-25G
Diethyl (4-methylbenzyl)phosphonate ≥98.0%
Diethyl (4-methylbenzyl)phosphonate

CAS Number: 3762-25-2
MDL Number: MFCD00041385
Molecular Formula: C12H19O3P
Molecular Weight: 242.25
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 160
Flash Point (°C): 110
Specific Gravity (20/20): 1.08


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Methylbenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₂H₁₉O₃P
MW: 242.26 g/mol
Boiling Pt: 160 °C
Density: 1.08 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041385
CAS Number: 3762-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44e987154636657f943a38b4b1d40ef68a831705&vid=44e987154636657f943a38b4b1d40ef68a831705&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR