Diethyl (4-chlorobenzyl)phosphonate ≥97.0%

Supplier: TCI America

Synonyms: Diethyl 4-chlorobenzylphosphonate

D3335-25G D3335-5G
TCD3335-25GEA 161.95 CAD
TCD3335-25G TCD3335-5G
Diethyl (4-chlorobenzyl)phosphonate ≥97.0%
Diethyl (4-chlorobenzyl)phosphonate

CAS Number: 39225-17-7
Molecular Formula: C11H16ClO3P
Molecular Weight: 262.67
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 110
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: (4-Chlorobenzyl)phosphonic Acid Diethyl Ester

Formula: C₁₁H₁₆ClO₃P
Boiling Pt: 180 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00051568
CAS Number: 39225-17-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65fe89a5d48e43e0c2f3b48c313075df6922f719&vid=65fe89a5d48e43e0c2f3b48c313075df6922f719&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR