3,4-Dimethylbenzaldehyde ≥95.0%

Supplier: TCI America

D3334-25G D3334-5G
TCD3334-25GEA 448.72 CAD
TCD3334-25G TCD3334-5G
3,4-Dimethylbenzaldehyde ≥95.0%
3,4-Dimethylbenzaldehyde

CAS Number: 5973-71-7
MDL Number: MFCD00016612
Molecular Formula: C9H10O
Molecular Weight: 134.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 226
Specific Gravity (20/20): 1.02


Other Product Names / Synonyms: o-Xylene-4-carboxaldehyde

Formula: C₉H₁₀O
Boiling Pt: 226 °C
Density: 1.02 (20 °C)
MDL Number: MFCD00016612
CAS Number: 5973-71-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d0cdb33d5e481dcd64a5e999c2000d3a541b310&vid=6ec1150fa4e887d888a3d955b9f63e0d26cd19ba&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR