2,3-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D3319-5G D3319-25G
TCD3319-5GEA 47.4 CAD
TCD3319-5G TCD3319-25G
2,3-Dichlorobenzoic acid ≥98.0% (by GC, titration analysis)
2,3-Dichlorobenzoic acid

CAS Number: 50-45-3
Molecular Formula: C7H4Cl2O2
Molecular Weight: 191.01
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169

Formula: C₇H₄Cl₂O₂
Melting Pt: 169 °C
MDL Number: MFCD00002413
CAS Number: 50-45-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a12898fc0335071a10398a321a15126da9bf0be3&vid=ed99fd78bc322afe163319c411b3755465dcbd82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR