3,5-Dichloro-2-fluoropyridine ≥98.0%

Supplier: TCI America

Synonyms: 2-Fluoro-3,5-dichloropyridine

D3317-1G D3317-5G
TCD3317-1GEA 36.34 CAD
TCD3317-1G TCD3317-5G
3,5-Dichloro-2-fluoropyridine ≥98.0%
3,5-Dichloro-2-fluoropyridine

CAS Number: 823-56-3
Molecular Formula: C5H2Cl2FN
Molecular Weight: 165.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 44

Formula: C₅H₂Cl₂FN
Melting Pt: 44 °C
MDL Number: MFCD04039349
CAS Number: 823-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=244850bb9ce16370dcaa8dd91579d7a0e1a35ca3&vid=244850bb9ce16370dcaa8dd91579d7a0e1a35ca3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR