5,6-Dimethoxy-1-indanone ≥98.0%
Supplier: TCI America
Synonyms:
5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one
D3313-25G
D3313-5G
TCD3313-25GEA
256.28
CAD
TCD3313-25G
TCD3313-5G
5,6-Dimethoxy-1-indanone ≥98.0%
5,6-Dimethoxy-1-indanone
CAS Number: 2107-69-9
Molecular Formula: C11H12O3
Molecular Weight: 192.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 139
Melting point (°C): 120
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 139 °C Melting Pt: 120 °C |
MDL Number:
MFCD00003790 CAS Number: 2107-69-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...