3,5-Dichloro-2-pyridone ≥98.0%

Supplier: TCI America

Synonyms: 3,5-Dichloro-2-hydroxypyridine

D3312-25G D3312-5G
TCD3312-25GEA 530.56 CAD
TCD3312-25G TCD3312-5G
3,5-Dichloro-2-pyridone ≥98.0%
3,5-Dichloro-2-pyridone

CAS Number: 5437-33-2
Molecular Formula: C5H3Cl2NO
Molecular Weight: 163.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 180


Other Product Names / Synonyms: 3,5-Dichloro-2-hydroxypyridine, 3,5-Dichloro-2-pyridinol

Formula: C₅H₃Cl₂NO
MW: 163.99 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00015401
CAS Number: 5437-33-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0d768cebf803cc2f6aea07c695c1afdc0e28760a&vid=0d768cebf803cc2f6aea07c695c1afdc0e28760a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR