Direct red 80
Supplier: TCI America
Synonyms:
CI 35780
, Sirius Red
, Sirius Red F3B
, Picro-Sirius Red
D3273-25G
TCD3273-25GEA
99.79
CAD
TCD3273-25G
Direct red 80
Direct Red 80
CAS Number: 2610-10-8
MDL Number: MFCD00054389
Molecular Formula: C45H32N10O21S6
Molecular Weight: 1373.05
Form: Crystal
Color: Deep Yellow Red
Other Product Names / Synonyms: Sirius Red
Formula:
C₄₅H₂₆N₁₀Na₆O₂₁S₆ MW: 1373.051 g/mol |
MDL Number:
MFCD00054389 CAS Number: 2610-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...