3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98.0%
Supplier: TCI America
Synonyms:
Diethyl squarate, 3,4-Diethoxycyclobut-3-ene-1,2-dione
D3253-25G
D3253-5G
TCD3253-25GEA
497.7
CAD
TCD3253-25G
TCD3253-5G
3,4-Diethoxy-3-cyclobutene-1,2-dione ≥98.0%
3,4-Diethoxy-3-cyclobutene-1,2-dione
CAS Number: 5231-87-8
Molecular Formula: C8H10O4
Molecular Weight: 170.16
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 95
Flash Point (°C): 110
Specific Gravity (20/20): 1.16
Other Product Names / Synonyms: Diethyl Squarate, Squaric Acid Diethyl Ester
Formula:
C₈H₁₀O₄ MW: 170.16 g/mol Boiling Pt: 95 °C Density: 1.16 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00001333 CAS Number: 5231-87-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...