2-(Diphenylphosphino)benzoic acid ≥98.0%

Supplier: TCI America

D3242-1G D3242-5G
TCD3242-1GEA 74.26 CAD
TCD3242-1G 76219-022
2-(Diphenylphosphino)benzoic acid ≥98.0%
2-(Diphenylphosphino)benzoic acid

CAS Number: 17261-28-8
Molecular Formula: C19H15O2P
Molecular Weight: 306.30
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 180

Formula: C₁₉H₁₅O₂P
MW: 306.3
Melting Pt: 180 °C
MDL Number: MFCD00674024
CAS Number: 17261-28-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ef1e5d4538fd0e1f9bb6294e188a7c9759497ae4&vid=ef1e5d4538fd0e1f9bb6294e188a7c9759497ae4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR