1,3-Dimethylimidazolium dimethylphosphate ≥97.0%

Supplier: TCI America

Synonyms: 1,3-Dimethylimidazolium Dimethyl Phosphate

D3240-25G D3240-5G
TCD3240-25GEA 143.78 CAD
TCD3240-25G TCD3240-5G
1,3-Dimethylimidazolium dimethylphosphate ≥97.0%
1,3-Dimethylimidazolium dimethylphosphate

CAS Number: 654058-04-5
Molecular Formula: C7H15N2O4P
Molecular Weight: 222.18
Purity/Analysis Method: >97.0% (N)
Form: Clear Liquid
Specific Gravity (20/20): 1.26

Formula: C₇H₁₅N₂O₄P
Density: 1.26 (20 °C)
MDL Number: MFCD08275369
CAS Number: 654058-04-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c6ec06c38142144683dcb3fe59305adb1b1dbe2&vid=2c6ec06c38142144683dcb3fe59305adb1b1dbe2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR