3,4-Dimethoxyphenol ≥97.0%

Supplier: TCI America

D3221-25G D3221-5G
TCD3221-25GEA 522.98 CAD
TCD3221-25G TCD3221-5G
3,4-Dimethoxyphenol ≥97.0%
3,4-Dimethoxyphenol

CAS Number: 2033-89-8
Molecular Formula: C8H10O3
Molecular Weight: 154.17
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 167
Melting point (°C): 80

Formula: C₈H₁₀O₃
Boiling Pt: 167 °C
Melting Pt: 80 °C
MDL Number: MFCD00008390
CAS Number: 2033-89-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e62a28a996a756fa75e86447fed659ee33529f95&vid=0c63597d3b3eedeadedeef7635a10451e22ad43d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR