3,6-Diphenyl-1,2,4,5-tetrazine ≥98.0% (by total nitrogen basis)
Supplier: TCI America
D3175-5G
D3175-1G
TCD3175-5GEA
355.5
CAD
TCD3175-5G
TCD3175-1G
3,6-Diphenyl-1,2,4,5-tetrazine ≥98.0% (by total nitrogen basis)
3,6-Diphenyl-1,2,4,5-tetrazine
CAS Number: 6830-78-0
Molecular Formula: C14H10N4
Molecular Weight: 234.26
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 196
Formula:
C₁₄H₁₀N₄ Melting Pt: 196 °C |
MDL Number:
MFCD00023646 CAS Number: 6830-78-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...