2,6-Difluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

Synonyms: Dihydroxy(2,6-difluorophenyl)borane, 2,6-Difluorobenzeneboronic acid, (2,6-Difluorophenyl)metaboric acid

D3087-1G D3087-5G D3087-25G
TCD3087-1GEA 39.5 CAD
TCD3087-1G TCD3087-5G 76219-016
2,6-Difluorophenylboronic acid (contains varying amounts of Anhydride)
2,6-Difluorophenylboronic acid

CAS Number: 162101-25-9
Molecular Formula: C6H5BF2O2
Molecular Weight: 157.91
Form: Crystal
Color: White
Melting point (°C): 149


Other Product Names / Synonyms: 2,6-Difluorobenzeneboronic Acid

Formula: C₆H₅BF₂O₂
MW: 157.91 g/mol
Melting Pt: 149 °C
MDL Number: MFCD00792436
CAS Number: 162101-25-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9039e080c1c1b5fd0464a5bd4b73450d2d60b03c&vid=9039e080c1c1b5fd0464a5bd4b73450d2d60b03c&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR