3,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
(3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid
D3081-5G
D3081-25G
TCD3081-5GEA
34.76
CAD
TCD3081-5G
TCD3081-25G
3,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
3,5-Difluorophenylboronic acid
CAS Number: 156545-07-2
MDL Number: MFCD01318138
Molecular Formula: C6H5BF2O2
Molecular Weight: 157.91
Form: Crystal
Color: White
Melting point (°C): 250
Other Product Names / Synonyms: 3,5-Difluorobenzeneboronic Acid
Formula:
C₆H₅BF₂O₂ MW: 157.91 g/mol Melting Pt: 250 °C |
MDL Number:
MFCD01318138 CAS Number: 156545-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...