2,3-Dichloropyrazine ≥98.0%
Supplier: TCI America
D3061-5G
D3061-25G
TCD3061-5GEA
48.24
CAD
TCD3061-5G
TCD3061-25G
2,3-Dichloropyrazine ≥98.0%
2,3-Dichloropyrazine
CAS Number: 4858-85-9
Molecular Formula: C4H2Cl2N2
Molecular Weight: 148.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 107
Flash Point (°C): 102
Freezing point (°C): 29
Formula:
C₄H₂Cl₂N₂ Boiling Pt: 107 °C Flash Pt: 102 °C |
MDL Number:
MFCD00040964 CAS Number: 4858-85-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...