2,5-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
D3044-5G
D3044-25G
TCD3044-5GEA
150.42
CAD
TCD3044-5G
TCD3044-25G
2,5-Dichlorophenylboronic acid (contains varying amounts of Anhydride)
2,5-Dichlorophenylboronic acid
CAS Number: 135145-90-3
MDL Number: MFCD01863182
Molecular Formula: C6H5BCl2O2
Molecular Weight: 190.81
Form: Crystal
Color: White
Other Product Names / Synonyms: 2,5-Dichlorobenzeneboronic Acid
Formula:
C₆H₅BCl₂O₂ |
MDL Number:
MFCD01863182 CAS Number: 135145-90-3 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...