(3,5-Dimethoxyphenyl)acetic acid ≥98.0%

Supplier: TCI America

Synonyms: 2-(3,5-Dimethoxyphenyl)acetic acid

D2950-1G D2950-5G
TCD2950-1GEA 55.3 CAD
TCD2950-1G TCD2950-5G
(3,5-Dimethoxyphenyl)acetic acid ≥98.0%
(3,5-Dimethoxyphenyl)acetic acid

CAS Number: 4670-10-4
MDL Number: MFCD00016827
Molecular Formula: C10H12O4
Molecular Weight: 196.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 104

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Melting Pt: 104 °C
MDL Number: MFCD00016827
CAS Number: 4670-10-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf719aa888cd20f1ff5cc938e4b46f26482e4d82&vid=cf719aa888cd20f1ff5cc938e4b46f26482e4d82&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR