2,5-Dihydroxybenzoic acid ≥99.0% (by GC, titration analysis)

Supplier: TCI America

D2933-5G D2933-1G
TCD2933-5GEA 124.84 CAD
TCD2933-5G 76219-014
2,5-Dihydroxybenzoic acid ≥99.0% (by GC, titration analysis)
2,5-Dihydroxybenzoic acid

[Matrix for MALDI-TOF/MS]
CAS Number: 490-79-9
MDL Number: MFCD00002460
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: >99.0% (GC,T)
Form: Crystal
Melting point (°C): 206


Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Gentisic Acid

Formula: C₇H₆O₄
Melting Pt: 206 °C
MDL Number: MFCD00002460
CAS Number: 490-79-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b91c98654769c073aa7ce10a3769ba31210ee2c1&vid=f27ec5252c63e9563d6a11ee928a168dfa818d3c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR