2,3-Dibromopyridine ≥98.0%
Supplier: TCI America
D2917-1G
D2917-5G
TCD2917-1GEA
26.86
CAD
TCD2917-1G
TCD2917-5G
2,3-Dibromopyridine ≥98.0%
2,3-Dibromopyridine
CAS Number: 13534-89-9
MDL Number: MFCD00234014
Molecular Formula: C5H3Br2N
Molecular Weight: 236.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 249
Melting point (°C): 59
Formula:
C₅H₃Br₂N Boiling Pt: 249 °C Melting Pt: 59 °C |
MDL Number:
MFCD00234014 CAS Number: 13534-89-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...