2,3-Dichloropyridine ≥98.0%

Supplier: TCI America

D2906-25G D2906-5G
TCD2906-25GEA 224.36 CAD
TCD2906-25G TCD2906-5G
2,3-Dichloropyridine ≥98.0%
2,3-Dichloropyridine

CAS Number: 2402-77-9
MDL Number: MFCD00006229
Molecular Formula: C5H3Cl2N
Molecular Weight: 147.99
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₅H₃Cl₂N
MW: 147.99 g/mol
Melting Pt: 68 °C
MDL Number: MFCD00006229
CAS Number: 2402-77-9
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da1508c2f87eae1adf950e03a7dc283c9cd2ee80&vid=da1508c2f87eae1adf950e03a7dc283c9cd2ee80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR