4-Nitro-2,3-lutidine-N-oxide ≥98.0%

Supplier: TCI America

Synonyms: 2,3-Dimethyl-4-nitropyridine 1-oxide, 2,3-Dimethyl-4-nitropyridine N-oxide

D2905-25G D2905-5G
TCD2905-25GEA 85.32 CAD
TCD2905-25G TCD2905-5G
4-Nitro-2,3-lutidine-N-oxide ≥98.0%
4-Nitro-2,3-lutidine N-oxide

CAS Number: 37699-43-7
MDL Number: MFCD00065172
Molecular Formula: C7H8N2O3
Molecular Weight: 168.15
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 92


Other Product Names / Synonyms: 4-Nitro-2,3-lutidine N-Oxide

Formula: C₇H₈N₂O₂
MW: 168.15 g/mol
Melting Pt: 92 °C
MDL Number: MFCD00065172
CAS Number: 37699-43-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=310c4bb6d42bb1b22ed004ecc311f9a1cd2cc105&vid=310c4bb6d42bb1b22ed004ecc311f9a1cd2cc105&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR