Methyl (diethylphosphono)acetate ≥96.0%

Supplier: TCI America

Synonyms: Diethyl methoxycarbonylmethanephosphonate, Methyl 2-(diethoxyphosphoryl)acetate, Methyl diethylphosphonoacetate

D2873-25G D2873-5G
TCD2873-25GEA 75.84 CAD
TCD2873-25G TCD2873-5G
Methyl (diethylphosphono)acetate ≥96.0%
Methyl (diethylphosphono)acetate

CAS Number: 1067-74-9
MDL Number: MFCD00009081
Molecular Formula: C7H15O5P
Molecular Weight: 210.17
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Product Categories: Synthetic Organic Chemistry, Carbonyl Olefination
Other Product Names / Synonyms: Diethylphosphonoacetic Acid Methyl Ester

Formula: C₇H₁₅O₅P
MW: 210.17 g/mol
Boiling Pt: 129 °C
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00009081
CAS Number: 1067-74-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a802c04ece94abe50ed7de499a92eaf6b8f0311&vid=7a802c04ece94abe50ed7de499a92eaf6b8f0311&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR