Methyl-3,5-dibromo-4-methylbenzoate ≥98.0%

Supplier: TCI America

D2871-25G D2871-5G
TCD2871-25GEA 237 CAD
TCD2871-25G TCD2871-5G
Methyl-3,5-dibromo-4-methylbenzoate ≥98.0%
Methyl 3,5-dibromo-4-methylbenzoate

CAS Number: 74896-66-5
MDL Number: MFCD00010729
Molecular Formula: C9H8Br2O2
Molecular Weight: 307.97
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87


Other Product Names / Synonyms: 3,5-Dibromo-p-toluic Acid Methyl Ester, 3,5-Dibromo-4-methylbenzoic Acid Methyl Ester, Methyl 3,5-Dibromo-p-toluate

Formula: C₉H₈Br₂O₂
Melting Pt: 87 °C
MDL Number: MFCD00010729
CAS Number: 74896-66-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4371f704b697624207d91a067bbb239d3eada6b&vid=d4371f704b697624207d91a067bbb239d3eada6b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR