4-Dodecyloxybenzoic acid ≥98.0%

Supplier: TCI America

Synonyms: 4-(Dodecyloxy)benzoic acid

D2859-25G D2859-5G
TCD2859-25GEA 267.02 CAD
TCD2859-25G TCD2859-5G
4-Dodecyloxybenzoic acid ≥98.0%
4-Dodecyloxybenzoic acid

CAS Number: 2312-15-4
MDL Number: MFCD00002543
Molecular Formula: C19H30O3
Molecular Weight: 306.45
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 137

Formula: C₁₉H₃₀O₃
Melting Pt: 137 °C
MDL Number: MFCD00002543
CAS Number: 2312-15-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7644d92a5266398fd90222a510798b35998fecf1&vid=1dd5bb7be28850fc27b0b61593684658cb3236ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR