1,2-Dibromo-4-fluorobenzene ≥98.0%
Supplier: TCI America
D2854-5G
D2854-25G
TCD2854-5GEA
60.04
CAD
TCD2854-5G
TCD2854-25G
1,2-Dibromo-4-fluorobenzene ≥98.0%
1,2-Dibromo-4-fluorobenzene
CAS Number: 2369-37-1
MDL Number: MFCD00070780
Molecular Formula: C6H3Br2F
Molecular Weight: 253.90
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Formula:
C₆H₃Br₂F MW: 253.9 |
MDL Number:
MFCD00070780 CAS Number: 2369-37-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...