Ethyl-3-(dimethylamino)acrylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D2836-25G D2836-100G
TCD2836-25GEA 121.66 CAD
TCD2836-25G TCD2836-100G
Ethyl-3-(dimethylamino)acrylate ≥98.0% (by GC, titration analysis)
Ethyl-3-(dimethylamino)acrylate

CAS Number: 924-99-2
MDL Number: MFCD00144269
Molecular Formula: C7H13NO2
Molecular Weight: 143.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 136
Melting point (°C): 19
Flash Point (°C): 108
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: 3-(Dimethylamino)acrylic Acid Ethyl Ester

Formula: C₇H₁₃NO₂
Boiling Pt: 136 °C
Melting Pt: 19 °C
Density: 1.00 (20 °C)
Flash Pt: 108 °C
MDL Number: MFCD00144269
CAS Number: 924-99-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57f5e40f7d62e9047e28896eb442fd80c61378f5&vid=57f5e40f7d62e9047e28896eb442fd80c61378f5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR