1,1'-Dibromoferrocene ≥98.0%

Supplier: TCI America

D2829-1G D2829-5G
TCD2829-1GEA 200.66 CAD
TCD2829-1G TCD2829-5G
1,1'-Dibromoferrocene ≥98.0%
1,1'-Dibromoferrocene

CAS Number: 1293-65-8
MDL Number: MFCD01075755
Molecular Formula: C10H8Br2Fe
Molecular Weight: 343.83
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C

Formula: C₁₀Br₂Fe
MDL Number: MFCD01075755
CAS Number: 1293-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c6ea013183be6760edad20b241aa82e7e4a46807&vid=c6ea013183be6760edad20b241aa82e7e4a46807&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR