Diphenylphosphine oxide ≥98.0%

Supplier: TCI America

D2826-25G D2826-5G
TCD2826-25GEA 793.16 CAD
TCD2826-25G TCD2826-5G
Diphenylphosphine oxide ≥98.0%
Diphenylphosphine oxide

CAS Number: 4559-70-0
MDL Number: MFCD00002079
Molecular Formula: C12H11OP
Molecular Weight: 202.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 105

Formula: C₁₂H₁₁OP
MW: 202.20 g/mol
Boiling Pt: 105 °C
MDL Number: MFCD00002079
CAS Number: 4559-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3187317474ec552b6f7ea058e681b21185f35917&vid=3187317474ec552b6f7ea058e681b21185f35917&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR