1,4-Diphenylbutadiyne ≥98.0%

Supplier: TCI America

D2789-25G D2789-5G
TCD2789-25GEA 601.66 CAD
TCD2789-25G TCD2789-5G
1,4-Diphenylbutadiyne ≥98.0%
1,4-Diphenylbutadiyne

CAS Number: 886-66-8
MDL Number: MFCD00004787
Molecular Formula: C16H10
Molecular Weight: 202.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 87

Formula: (C₆H₅)CCCC(C₆H₅)
MW: 202.26 g/mol
Boiling Pt: 210 °C
Melting Pt: 87 °C
MDL Number: MFCD00004787
CAS Number: 886-66-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e551d9d47ed60871fb9d4bb9176ce4d1863476b6&vid=e551d9d47ed60871fb9d4bb9176ce4d1863476b6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR