1-Bromo-3,3-diphenylpropane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(3-Bromopropane-1,1-diyl)dibenzene, 1,1'-(3-Bromopropylidene)bisbenzene
D2784-25G
D2784-5G
TCD2784-25GEA
1122.27
CAD
TCD2784-25G
TCD2784-5G
1-Bromo-3,3-diphenylpropane ≥98.0% (by GC, titration analysis)
1-Bromo-3,3-diphenylpropane
CAS Number: 20017-68-9
MDL Number: MFCD00044829
Molecular Formula: C15H15Br
Molecular Weight: 275.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 40
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 3-Bromo-1,1-diphenylpropane
Formula:
(C₆H₅)₂CHCH₂CH₂Br MW: 275.19 g/mol Boiling Pt: 145 °C Melting Pt: 40 °C |
MDL Number:
MFCD00044829 CAS Number: 20017-68-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...