2,6-Dimethoxybenzaldehyde ≥97.0%
Supplier: TCI America
D2778-25G
D2778-5G
TCD2778-25GEA
619.36
CAD
TCD2778-25G
TCD2778-5G
2,6-Dimethoxybenzaldehyde ≥97.0%
2,6-Dimethoxybenzaldehyde
CAS Number: 3392-97-0
MDL Number: MFCD00010862
Molecular Formula: C9H10O3
Molecular Weight: 166.18
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 97
Formula:
C₉H₁₀O₃ Boiling Pt: 158 °C Melting Pt: 97 °C |
MDL Number:
MFCD00010862 CAS Number: 3392-97-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...