4,5-Dichloro-3(2H)-pyridazinone 98,0

Supplier: TCI America

Synonyms: 4,5-Dichloro-3-hydroxypyridazine, 4,5-dichloro-2,3-dihydropyridazin-3-one

D2768-100G D2768-25G
TCD2768-100GEA 325.48 CAD
TCD2768-100G TCD2768-25G
4,5-Dichloro-3(2H)-pyridazinone 98,0
4,5-Dichloro-3(2H)-pyridazinone

CAS Number: 932-22-9
MDL Number: MFCD00051504
Molecular Formula: C4H2Cl2N2O
Molecular Weight: 164.97
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 203


Other Product Names / Synonyms: 4,5-Dichloro-3-hydroxypyridazine

Formula: C₄H₂Cl₂N₂O
MW: 164.98 g/mol
Melting Pt: 203 °C
MDL Number: MFCD00051504
CAS Number: 932-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=30c703c41a9b4ddfd9619dd36f82443a89abac75&vid=30c703c41a9b4ddfd9619dd36f82443a89abac75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR