Diethyl dipropylmalonate ≥97.0%

Supplier: TCI America

D2762-25ML D2762-500ML
TCD2762-25MLEA 52.14 CAD
TCD2762-25ML TCD2762-500ML
Diethyl dipropylmalonate ≥97.0%
Diethyl dipropylmalonate

CAS Number: 6065-63-0
MDL Number: MFCD00026840
Molecular Formula: C13H24O4
Molecular Weight: 244.33
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 249
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: Dipropylmalonic Acid Diethyl Ester

Formula: C₁₃H₂₄O₄
Boiling Pt: 249 °C
Density: 0.96 (20 °C)
MDL Number: MFCD00026840
CAS Number: 6065-63-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a22ca367dffadeef4767e1a6333e45dc679c26e7&vid=a22ca367dffadeef4767e1a6333e45dc679c26e7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR