2,6-Dichloro-3-nitropyridine ≥98.0%

Supplier: TCI America

D2761-25G D2761-5G
TCD2761-25GEA 142.2 CAD
TCD2761-25G TCD2761-5G
2,6-Dichloro-3-nitropyridine ≥98.0%
2,6-Dichloro-3-nitropyridine

CAS Number: 16013-85-7
MDL Number: MFCD00006234
Molecular Formula: C5H2Cl2N2O2
Molecular Weight: 192.98
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 65
Flash Point (°C): 110


Other Product Names / Synonyms: 3-Nitro-2,6-dichloropyridine

Formula: C₅H₂Cl₂N₂O₂
Melting Pt: 65 °C
Flash Pt: 110 °C
MDL Number: MFCD00006234
CAS Number: 16013-85-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=84e7083f9e8f20d4394f852b9bd16c6b089cb1da&vid=84e7083f9e8f20d4394f852b9bd16c6b089cb1da&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR