Dichloro(p-cymene)ruthenium(II) dimer ≥95.0%

Supplier: TCI America

Synonyms: Di(μ-chloro)dichlorobis(p-cymene)diruthenium(II)

D2751-1G D2751-5G
TCD2751-1GEA 88.48 CAD
TCD2751-1G TCD2751-5G
Dichloro(p-cymene)ruthenium(II) dimer ≥95.0%
Dichloro(p-cymene)ruthenium(II) dimer

CAS Number: 52462-29-0
MDL Number: MFCD00064793
Molecular Formula: C20H28Cl4Ru2
Molecular Weight: 612.38
Purity/Analysis Method: >95.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Activation Reaction of Unactivated Bonds, C-H Bond Activation Reaction
Other Product Names / Synonyms: p-Cymeneruthenium(II) Dichloride Dimer, Di-mu-chloro-bis[chloro(p-cymene)ruthenium(II)]

Formula: C₂₀H₂₈Cl₄Ru₂
MW: 612.38 g/mol
MDL Number: MFCD00064793
CAS Number: 52462-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b9194d6831fc6400224f7c78dd07339a8f21d318&vid=b9194d6831fc6400224f7c78dd07339a8f21d318&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR