2',6'-Dichloro-3'-fluoroacetophenone ≥97.0%
Supplier: TCI America
D2750-25G
D2750-5G
TCD2750-25GEA
128.93
CAD
TCD2750-25G
TCD2750-5G
2',6'-Dichloro-3'-fluoroacetophenone ≥97.0%
2',6'-Dichloro-3'-fluoroacetophenone
CAS Number: 290835-85-7
MDL Number: MFCD02093760
Molecular Formula: C8H5Cl2FO
Molecular Weight: 207.03
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Specific Gravity (20/20): 1.40
Formula:
C₈H₅Cl₂FO MW: 207.03 g/mol Boiling Pt: 255 °C Density: 1.40 (20 °C) |
MDL Number:
MFCD02093760 CAS Number: 290835-85-7 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...