(S)-(-)-α,α-Diphenylprolinol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(S)-(-)-α,α-Diphenyl-2-pyrrolidinemethanol
CAS Number: 112068-01-6
MDL Number: MFCD00075506
Molecular Formula: C17H19NO
Molecular Weight: 253.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79
Specific rotation [a]20/D: -66.5 deg (C=3, CHCl3)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-(-)-2-(Diphenylhydroxymethyl)pyrrolidine, alpha,alpha-Diphenyl-L-prolinol
Formula:
C₁₇H₁₉NO MW: 253.35 Melting Pt: 79 °C |
MDL Number:
MFCD00075506 CAS Number: 112068-01-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...