2,5-Dichloro-4-nitroaniline ≥98.0%

Supplier: TCI America

D2722-25G D2722-5G
TCD2722-25GEA 475.58 CAD
TCD2722-25G TCD2722-5G
2,5-Dichloro-4-nitroaniline ≥98.0%
2,5-Dichloro-4-nitroaniline

CAS Number: 6627-34-5
MDL Number: MFCD00025157
Molecular Formula: C6H4Cl2N2O2
Molecular Weight: 207.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 155

Formula: C₆H₄Cl₂N₂O₂
MW: 207.02 g/mol
Melting Pt: 155 °C
MDL Number: MFCD00025157
CAS Number: 6627-34-5
UN: 2237
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91287bc49eff2175e00e07005a4e609f3e8cab36&vid=255978502f8c62885073d39bdafc42fe3f6b0a0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR