Methyl-2,5-dibromobenzoate ≥98.0%
Supplier: TCI America
D2673-5G
D2673-25G
TCD2673-005GEA
72.68
CAD
TCD2673-005G
TCD2673-025G
Methyl-2,5-dibromobenzoate ≥98.0%
Methyl 2,5-dibromobenzoate
CAS Number: 57381-43-8
MDL Number: MFCD00144757
Molecular Formula: C8H6Br2O2
Molecular Weight: 293.94
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50
Other Product Names / Synonyms: 2,5-Dibromobenzoic Acid Methyl Ester
Formula:
C₈H₆Br₂O₂ Melting Pt: 50 °C |
MDL Number:
MFCD00144757 CAS Number: 57381-43-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...