Dichloro(1,5-cyclooctadiene)palladium(II) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
1,5-Cyclooctadienepalladium(II) Dichloride
D2604-5G
D2604-1G
TCD2604-5GEA
1993.96
CAD
TCD2604-5G
TCD2604-001G
Dichloro(1,5-cyclooctadiene)palladium(II) ≥98.0% (by titrimetric analysis)
Dichloro(1,5-cyclooctadiene)palladium(II)
CAS Number: 12107-56-1
MDL Number: MFCD00012412
Molecular Formula: C8H12Cl2Pd
Molecular Weight: 285.50
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 210
Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride
Formula:
C₈H₁₂Cl₂Pd MW: 285.50 g/mol Melting Pt: 210 °C |
MDL Number:
MFCD00012412 CAS Number: 12107-56-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...