Ethoxydiphenylphosphine ≥93.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Diphenylphosphinous Acid Ethyl Ester
D2558-25G
D2558-5G
TCD2558-25GEA
373.51
CAD
TCD2558-25G
TCD2558-005G
Ethoxydiphenylphosphine ≥93.0% (by titrimetric analysis)
Ethoxydiphenylphosphine
CAS Number: 719-80-2
MDL Number: MFCD00009087
Molecular Formula: C14H15OP
Molecular Weight: 230.25
Purity/Analysis Method: >93.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.07
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Diphenylphosphinous Acid Ethyl Ester
Formula:
C₁₄H₁₅OP MW: 230.25 g/mol Density: 1.07 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009087 CAS Number: 719-80-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...