(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine ≥97.0%

Supplier: TCI America

D2529-200MG D2529-1G
TCD2529-200MGEA 105.86 CAD
TCD2529-200MG TCD2529-001G
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine ≥97.0%
(S)-(-)-N,N-Dimethyl-1-ferrocenylethylamine

CAS Number: 31886-57-4
MDL Number: MFCD00066272
Molecular Formula: C14H19FeN
Molecular Weight: 257.16
Purity/Analysis Method: >97.0% (GC)
Form: Liquid
Boiling point (°C): 121
Flash Point (°C): 95
Specific Gravity (20/20): 1.22
Specific rotation [a]20/D: -14 deg (C=1, EtOH)


Other Product Names / Synonyms: (S)-(-)-[1-(Dimethylamino)ethyl]ferrocene

Formula: C₁₄H₁₉FeN
Boiling Pt: 121 °C
Density: 1.22 (20 °C)
Flash Pt: 95 °C
MDL Number: MFCD00066272
CAS Number: 31886-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=305a095b983e250693a236573faf949fd3586ae0&vid=305a095b983e250693a236573faf949fd3586ae0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR