(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0%

Supplier: TCI America

D2528-200MG D2528-1G
TCD2528-200MGEA 140.62 CAD
TCD2528-200MG TCD2528-001G
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine ≥98.0%
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine

CAS Number: 31886-58-5
MDL Number: MFCD00066273
Molecular Formula: C14H19FeN
Molecular Weight: 257.16
Purity/Analysis Method: >98.0% (GC)
Form: Liquid
Boiling point (°C): 93
Flash Point (°C): 110
Specific Gravity (20/20): 1.22
Specific rotation [a]20/D: 15 deg (C=1, EtOH)


Other Product Names / Synonyms: (R)-(+)-[1-(Dimethylamino)ethyl]ferrocene

Formula: C₁₄H₁₉FeN
Boiling Pt: 93 °C
Density: 1.22 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00066273
CAS Number: 31886-58-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69127a98c9d04a237114ce9fb84cc81b2854537e&vid=69127a98c9d04a237114ce9fb84cc81b2854537e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR