1,1-Diphenyl-2-propyn-1-ol ≥97.0%

Supplier: TCI America

D2495-25G D2495-5G
TCD2495-25GEA 309.05 CAD
TCD2495-25G TCD2495-005G
1,1-Diphenyl-2-propyn-1-ol ≥97.0%
1,1-Diphenyl-2-propyn-1-ol

CAS Number: 3923-52-2
MDL Number: MFCD00041570
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 200
Melting point (°C): 48

Formula: C₁₅H₁₂O
Boiling Pt: 200 °C
Melting Pt: 48 °C
MDL Number: MFCD00041570
CAS Number: 3923-52-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ec2ee2d8e1ec9215bc614337cfb99d93b3a30037&vid=ec2ee2d8e1ec9215bc614337cfb99d93b3a30037&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR