4,6-Dimethyl-2-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-4,6-dimethylpyrimidine, 4,6-Dimethyl-2-pyrimidinol, 4,6-Dimethylpyrimidin-2(1H)-one

D2427-500G D2427-25G D2427-100G
TCD2427-500GEA 922.72 CAD
TCD2427-500G TCD2427-025G TCD2427-100G
4,6-Dimethyl-2-hydroxypyrimidine ≥98.0% (by HPLC, titration analysis)
4,6-Dimethyl-2-hydroxypyrimidine

CAS Number: 108-79-2
MDL Number: MFCD00006072
Molecular Formula: C6H8N2O
Molecular Weight: 124.14
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 202


Other Product Names / Synonyms: 4,6-Dimethyl-2-pyrimidinol, 2-Hydroxy-4,6-dimethylpyrimidine

Formula: C₆H₈N₂O
MW: 124.14 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00006072
CAS Number: 108-79-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74e9b4ecd5a38eec2bbf386ca27d2d5f68d4dfe7&vid=74e9b4ecd5a38eec2bbf386ca27d2d5f68d4dfe7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR